3,6-Difluoro-2-hydroxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

17258-5G 17258-1G 17258-25G 17258-500MG
77627-928EA 1484.23 USD
77627-928 77636-948 77653-874 77644-934
3,6-Difluoro-2-hydroxyphenylboronic acid ≥95%
3,6-Difluoro-2-hydroxyphenylboronic acid

Precaución: For Research Use Only

Formula: C₆H₅BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD18394882
CAS Number: 2246616-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR