6-Bromo-2,4-difluoro-3-methylphenol ≥97%

Supplier: AOBChem USA

Warning

172438-5G 172438-1G 172438-500MG
77627-926EA 1688.17 USD
77627-926 77640-942 77648-962
6-Bromo-2,4-difluoro-3-methylphenol ≥97%
6-Bromo-2,4-difluoro-3-methylphenol

Precaución: For Research Use Only

Formula: C₇H₅BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD33411680
CAS Number: 1784551-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR