2,4-Difluoro-α,α,3-trimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

172434-1G 172434-5G 172434-500MG
77636-946EA 666 USD
77636-946 77653-872 77644-932
2,4-Difluoro-α,α,3-trimethylbenzenemethanol ≥95%
2,4-Difluoro-α,α,3-trimethylbenzenemethanol

Precaución: For Research Use Only

Formula: 186.2 g/mol
MW: C₁₀H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD28522164
CAS Number: 1785572-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR