5-Iodothiophene-2-carbonitrile ≥97%

Supplier: AOBChem USA

Warning

172136-5G 172136-25G 172136-1G 172136-500MG
77631-818EA 1053 USD
77631-818 77653-868 77640-938 77644-928
5-Iodothiophene-2-carbonitrile ≥97%
5-Iodothiophene-2-carbonitrile

Precaución: For Research Use Only

Formula: C₅H₂INS
MW: 235.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14584473
CAS Number: 18945-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR