2-Acetyl-3-chloro-5-(trifluoromethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-2-ethanoyl-5-(trifluoromethyl)pyridine, 1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]ethan-1-one

172004-5G 172004-25G 172004-1G 172004-500MG
77631-816EA 642 USD
77631-816 77657-720 77644-926 77648-960
2-Acetyl-3-chloro-5-(trifluoromethyl)pyridine ≥95%
2-Acetyl-3-chloro-5-(trifluoromethyl)pyridine

Precaución: For Research Use Only

Formula: C₈H₅ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD11113406
CAS Number: 207994-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR