5-Bromo-2-fluoro-3-(trifluoromethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

171964-5G 171964-25G 171964-1G 171964-500MG
77631-814EA 989 USD
77631-814 77653-866 77640-936 77644-924
5-Bromo-2-fluoro-3-(trifluoromethyl)pyridine ≥95%
5-Bromo-2-fluoro-3-(trifluoromethyl)pyridine

Precaución: For Research Use Only

Formula: C₆H₂BrF₄N
MW: 243.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28675294
CAS Number: 1365262-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR