6-Methoxy-5-(trifluoromethyl)nicotinaldehyde ≥95%
Supplier: AOBChem USA
|
Warning
|
171790-5G
171790-25G
171790-1G
171790-500MG
77634-188EA
434
USD
77634-188
77653-864
77640-934
77644-922
6-Methoxy-5-(trifluoromethyl)nicotinaldehyde ≥95%
6-Methoxy-5-(trifluoromethyl)nicotinaldehyde
Precaución: For Research Use Only
Formula:
C₈H₆F₃NO₂ MW: 205.1340 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18254532 CAS Number: 1211519-38-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...