1-[6-Methoxy-5-(trifluoromethyl)-3-pyridinyl]ethanone ≥95%

Supplier: AOBChem USA

Warning

171788-5G 171788-1G 171788-500MG
77657-718EA 2445 USD
77657-718 77640-932 77648-958
1-[6-Methoxy-5-(trifluoromethyl)-3-pyridinyl]ethanone ≥95%
1-[6-Methoxy-5-(trifluoromethyl)-3-pyridinyl]ethanone

Precaución: For Research Use Only

Formula: 219.16 g/mol
MW: C₉H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD31567922
CAS Number: 1839581-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR