6-Methoxy-5-(trifluoromethyl)nicotinic acid ≥95%

Supplier: AOBChem USA

Warning

171781-5G 171781-25G 171781-1G 171781-500MG
77634-186EA 282 USD
77634-186 77653-862 77640-930 77648-956
6-Methoxy-5-(trifluoromethyl)nicotinic acid ≥95%
6-Methoxy-5-(trifluoromethyl)nicotinic acid

Precaución: For Research Use Only

Formula: C₈H₆F₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD18254533
CAS Number: 1211532-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR