3-Iodo-5-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

171774-5G 171774-25G 171774-1G 171774-500MG
77634-184EA 394 USD
77634-184 77653-860 77640-928 77648-954
3-Iodo-5-methoxypyridine ≥95%
3-Iodo-5-methoxypyridine

Precaución: For Research Use Only

Formula: C₆H₆INO
MW: 235.224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08235136
CAS Number: 873302-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR