3-Bromo-2,4-difluorobenzotrifluoride ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-1,3-difluoro-4-(trifluoromethyl)benzene

171633-5G 171633-1G 171633-500MG
77657-716EA 2274 USD
77657-716 77640-926 77644-920
3-Bromo-2,4-difluorobenzotrifluoride ≥95%
3-Bromo-2,4-difluorobenzotrifluoride

Precaución: For Research Use Only

Formula: C₇H₂BrF₅
MW: 260.987 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28737902
CAS Number: 1805023-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR