3-Chloro-2,4-difluorobenzotrifluoride ≥95%

Supplier: AOBChem USA

Warning

171617-5G 171617-25G 171617-1G 171617-500MG
77627-924EA 1755.12 USD
77627-924 77657-714 77640-924 77648-952
3-Chloro-2,4-difluorobenzotrifluoride ≥95%
3-Chloro-2,4-difluorobenzotrifluoride

Precaución: For Research Use Only

Formula: C₇H₂ClF₅
MW: 216.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972709
CAS Number: 120770-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR