1,3-Dichloro-2,5-difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

171455-5G 171455-25G
77634-178EA 143 USD
77634-178 77653-854
1,3-Dichloro-2,5-difluorobenzene ≥95%
1,3-Dichloro-2,5-difluorobenzene

Precaución: For Research Use Only

Formula: C₆H₂Cl₂F₂
MW: 182.983 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658263
CAS Number: 2367-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR