3-Bromo-2,6-dimethoxyphenol ≥97%

Supplier: AOBChem USA

Warning

17114-5G 17114-25G 17114-1G 17114-500MG
77631-810EA 803 USD
77631-810 77653-852 77640-918 77648-948
3-Bromo-2,6-dimethoxyphenol ≥97%
3-Bromo-2,6-dimethoxyphenol

Precaución: For Research Use Only

Formula: 233.059 g/mol
MW: C₈H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD28976907
CAS Number: 18111-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR