3-(Benzyloxy)-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

17112-5G 17112-1G 17112-25G 17112-500MG
77631-808EA 864 USD
77631-808 77636-942 77653-850 77648-946
3-(Benzyloxy)-2-fluorobenzoic acid ≥97%
3-(Benzyloxy)-2-fluorobenzoic acid

Precaución: For Research Use Only

Formula: C₁₄H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD28976906
CAS Number: 186584-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR