2-Bromo-6-fluoro-3-phenylmethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

17083-1G 17083-5G 17083-500MG
77636-940EA 525 USD
77636-940 77653-848 77644-914
2-Bromo-6-fluoro-3-phenylmethoxybenzaldehyde ≥97%
2-Bromo-6-fluoro-3-phenylmethoxybenzaldehyde

Precaución: For Research Use Only

Formula: C₁₄H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28976904
CAS Number: 2244107-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR