2,3-Difluoro-α,α-dimethyl-4-pyridinemethanol ≥95%
Supplier: AOBChem USA
|
Warning
|
170269-5G
170269-25G
170269-1G
170269-500MG
77627-918EA
1484.23
USD
77627-918
77635-482
77636-934
77648-940
2,3-Difluoro-α,α-dimethyl-4-pyridinemethanol ≥95%
2,3-Difluoro-α,α-dimethyl-4-pyridinemethanol
Precaución: For Research Use Only
Formula:
173.16 g/mol MW: C₈H₉F₂NO Storage Temperature: Ambient |
MDL Number:
MFCD34853091 CAS Number: 1636118-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...