1-(4-Fluorophenyl)butan-1-ol ≥95%

Supplier: AOBChem USA

Warning

170102-5G 170102-1G 170102-25G 170102-500MG
77631-804EA 998 USD
77631-804 77636-932 77653-844 77644-910
1-(4-Fluorophenyl)butan-1-ol ≥95%
1-(4-Fluorophenyl)butan-1-ol

Precaución: For Research Use Only

Formula: 168.2080 g/mol
MW: C₁₀H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD00046684
CAS Number: 704-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR