5-Bromo-3-fluoro-2-(phenylmethoxy)benzaldehyde, ≥97%

Supplier: AOBChem USA

Warning

17005-5G 17005-25G 17005-1G 17005-500MG
77631-802EA 803 USD
77631-802 77657-704 77640-914 77648-938
5-Bromo-3-fluoro-2-(phenylmethoxy)benzaldehyde, ≥97%
5-Bromo-3-fluoro-2-(phenylmethoxy)benzaldehyde,

Precaución: For Research Use Only

Formula: 309.13 g/mol
MW: C₁₄H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD12025064
CAS Number: 1003298-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR