4-(3-Bromophenyl)tetrahydro-2H-thiopyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

170011-5G 170011-25G 170011-1G 170011-500MG
77631-800EA 888 USD
77631-800 77653-842 77640-912 77648-936
4-(3-Bromophenyl)tetrahydro-2H-thiopyran-4-ol ≥97%
4-(3-Bromophenyl)tetrahydro-2H-thiopyran-4-ol

Precaución: For Research Use Only

Formula: 273.1893 g/mol
MW: C₁₁H₁₃BrOS
Storage Temperature: Ambient
MDL Number: MFCD34853017
CAS Number: 1781119-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR