4-(4-Bromophenyl)tetrahydro-2H-thiopyran ≥97%

Supplier: AOBChem USA

Warning

170010-5G 170010-1G 170010-500MG
77657-700EA 2023 USD
77657-700 77640-910 77644-908
4-(4-Bromophenyl)tetrahydro-2H-thiopyran ≥97%
4-(4-Bromophenyl)tetrahydro-2H-thiopyran

Precaución: For Research Use Only

Formula: 257.1899 g/mol
MW: C₁₁H₁₃BrS
Storage Temperature: Ambient
CAS Number: 1781345-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR