2,4-Dibromo-6-fluoroanisole ≥95%

Supplier: AOBChem USA

Warning

17001-5G 17001-25G
77634-174EA 178 USD
77634-174 77657-702
2,4-Dibromo-6-fluoroanisole ≥95%
2,4-Dibromo-6-fluoroanisole

Precaución: For Research Use Only

Formula: C₇H₅Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD00070788
CAS Number: 202982-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR