4-Bromo-3-(trifluoromethoxy)aniline ≥97%

Supplier: AOBChem USA

Warning

16980-5G 16980-1G 16980-25G
77634-170EA 227 USD
77634-170 77636-926 77657-694
4-Bromo-3-(trifluoromethoxy)aniline ≥97%
4-Bromo-3-(trifluoromethoxy)aniline

Precaución: For Research Use Only

Formula: C₇H₅BrF₃NO
MW: 256.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11506046
CAS Number: 116369-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR