4-Bromo-3-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16977-5G 16977-1G 16977-25G 16977-500MG
77631-792EA 699 USD
77631-792 77636-922 77653-836 77648-932
4-Bromo-3-(trifluoromethoxy)benzaldehyde ≥97%
4-Bromo-3-(trifluoromethoxy)benzaldehyde

Precaución: For Research Use Only

Formula: C₈H₄BrF₃O₂
MW: 269.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12924842
CAS Number: 1221716-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR