2-Chloro-6-methoxynaphthalene ≥97%

Supplier: AOBChem USA

Warning

169472-5G 169472-1G 169472-25G 169472-500MG
77631-788EA 1054 USD
77631-788 77636-912 77657-684 77644-898
2-Chloro-6-methoxynaphthalene ≥97%
2-Chloro-6-methoxynaphthalene

Precaución: For Research Use Only

Formula: C₁₁H₉ClO
MW: 192.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17012373
CAS Number: 67886-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR