4-Bromo-2-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

169399-5G 169399-1G 169399-500MG
77657-682EA 1869 USD
77657-682 77640-908 77644-896
4-Bromo-2-(methylthio)pyridine ≥95%

Precaución: For Research Use Only

Formula: C₆H₆BrNS
Storage Temperature: Ambient
MDL Number: MFCD11226871
CAS Number: 769163-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR