5-Bromo-3-methyl-N-methylpyridine-2-carboxamide ≥97%

Supplier: AOBChem USA

Warning

16937-5G 16937-1G 16937-25G 16937-500MG
77634-164EA 472 USD
77634-164 77636-908 77657-680 77648-922
5-Bromo-3-methyl-N-methylpyridine-2-carboxamide ≥97%
5-Bromo-3-methyl-N-methylpyridine-2-carboxamide

Precaución: For Research Use Only

Formula: 229.074 g/mol
MW: C₈H₉BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD11617268
CAS Number: 1224604-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR