2,5-Dibromo-4-pyridinylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(2,5-Dibromopyridin-4-yl)boronic acid
16934-5G
16934-1G
16934-25G
16934-500MG
77634-162EA
308
USD
77634-162
77636-906
77653-830
77648-920
2,5-Dibromo-4-pyridinylboronic acid ≥97%
2,5-Dibromo-4-pyridinylboronic acid
Precaución: For Research Use Only
Formula:
C₅H₄BBr₂NO₂ MW: 280.71 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11109338 CAS Number: 1031843-77-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...