6-Chloro-2-fluoro-3-(trimethylsilyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

169293-5G 169293-1G 169293-500MG
77627-916EA 1484.23 USD
77627-916 77636-902 77644-892
6-Chloro-2-fluoro-3-(trimethylsilyl)pyridine ≥95%
6-Chloro-2-fluoro-3-(trimethylsilyl)pyridine

Precaución: For Research Use Only

Formula: C₈H₁₁ClFNSi
MW: 203.72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32861402
CAS Number: 2639430-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR