2-Methoxy-5-(trimethylsilyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

169271-5G 169271-1G 169271-500MG
77627-914EA 1777.78 USD
77627-914 77644-890 77648-918
2-Methoxy-5-(trimethylsilyl)pyridine ≥95%
2-Methoxy-5-(trimethylsilyl)pyridine

Precaución: For Research Use Only

Formula: 181.31 g/mol
MW: C₉H₁₅NOSi
Storage Temperature: Ambient
MDL Number: MFCD32264541
CAS Number: 1197831-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR