4-Bromo-3-chloro-5-methyl-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

169227-5G 169227-25G 169227-1G 169227-500MG
77627-912EA 1484.23 USD
77627-912 77635-480 77640-906 77648-916
4-Bromo-3-chloro-5-methyl-1H-indazole ≥95%
4-Bromo-3-chloro-5-methyl-1H-indazole

Precaución: For Research Use Only

Formula: 245.5 g/mol
MW: C₈H₆BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD34852683
CAS Number: 2368910-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR