Methyl 5-formyl-4-methyl-2-thiophenecarboxylate ≥97%

Supplier: AOBChem USA

Warning

168841-5G 168841-1G 168841-500MG
77627-908EA 1484.23 USD
77627-908 77636-900 77644-886
Methyl 5-formyl-4-methyl-2-thiophenecarboxylate ≥97%
Methyl 5-formyl-4-methyl-2-thiophenecarboxylate

Precaución: For Research Use Only

Formula: C₈H₈O₃S
Storage Temperature: Ambient
MDL Number: MFCD04972647
CAS Number: 391936-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR