α,α,3,4-Tetramethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

168741-5G 168741-1G 168741-500MG
77653-828EA 1857 USD
77653-828 77640-900 77644-884
α,α,3,4-Tetramethylbenzenemethanol ≥95%
α,α,3,4-Tetramethylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₁H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD16158853
CAS Number: 40180-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR