2-(2,3-Dimethylphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

168730-5G 168730-1G 168730-500MG
77627-906EA 1484.23 USD
77627-906 77636-898 77644-882
2-(2,3-Dimethylphenyl)propan-2-ol ≥97%
2-(2,3-Dimethylphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₆O
MW: 164.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16794554
CAS Number: 156744-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR