2-Ethyl-α,α-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

168710-5G 168710-1G 168710-500MG
77657-670EA 1854 USD
77657-670 77640-898 77644-880
2-Ethyl-α,α-dimethylbenzenemethanol ≥95%
2-Ethyl-α,α-dimethylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₁H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD09965918
CAS Number: 104174-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR