3,5-Dibromo-1-benzyloxybenzene ≥97%

Supplier: AOBChem USA

Warning

16870-5G 16870-1G 16870-25G 16870-500MG
77631-782EA 827 USD
77631-782 77636-894 77657-668 77648-910
3,5-Dibromo-1-benzyloxybenzene ≥97%
3,5-Dibromo-1-benzyloxybenzene

Precaución: For Research Use Only

Formula: C₁₃H₁₀Br₂O
Storage Temperature: Ambient
MDL Number: MFCD17011975
CAS Number: 128924-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR