4-(Methoxymethyl)-α-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

168694-5G 168694-1G 168694-25G 168694-500MG
77631-780EA 1186 USD
77631-780 77636-892 77653-826 77648-908
4-(Methoxymethyl)-α-methylbenzenemethanol ≥95%
4-(Methoxymethyl)-α-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD21267811
CAS Number: 66190-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR