α-Methyl-4-(1-methylethoxy)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

168691-5G 168691-25G 168691-1G 168691-500MG
77631-778EA 1192 USD
77631-778 77657-664 77644-874 77644-876
α-Methyl-4-(1-methylethoxy)benzenemethanol ≥97%
α-Methyl-4-(1-methylethoxy)benzenemethanol

Precaución: For Research Use Only

Formula: 180.24 g/mol
MW: C₁₁H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD08443351
CAS Number: 586960-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR