1-(2,4-Dimethylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

168669-5G 168669-25G
77634-156EA 117 USD
77634-156 77653-824
1-(2,4-Dimethylphenyl)ethanol ≥95%
1-(2,4-Dimethylphenyl)ethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060890
CAS Number: 5379-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR