2-(4-Hydroxyphenyl)-2-propanol ≥95%

Supplier: AOBChem USA

Warning

168664-5G 168664-25G 168664-1G 168664-500MG
77634-154EA 493 USD
77634-154 77653-820 77640-894 77644-872
2-(4-Hydroxyphenyl)-2-propanol ≥95%
2-(4-Hydroxyphenyl)-2-propanol

Precaución: For Research Use Only

Formula: C₉H₁₂O₂
Storage Temperature: Ambient
CAS Number: 2948-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR