1-(3-Methylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: α,3-Dimethylbenzyl alcohol, m,α-Dimethylbenzyl alcohol, 3-(1-Hydroxyethyl)toluene, 3-Methylphenyl methyl carbinol, 1-(m-Tolyl)ethanol

168653-5G 168653-25G
77634-152EA 87 USD
77634-152 77653-818
1-(3-Methylphenyl)ethanol ≥95%
1-(3-Methylphenyl)ethanol

Precaución: For Research Use Only

Formula: C₉H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD00046635
CAS Number: 7287-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR