α,α-Dimethyl-2-(methylthio)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

168647-5G 168647-1G 168647-500MG
77653-816EA 2250 USD
77653-816 77640-892 77644-870
α,α-Dimethyl-2-(methylthio)benzenemethanol ≥95%
α,α-Dimethyl-2-(methylthio)benzenemethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD20528512
CAS Number: 6518-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR