3-Bromo-2,6-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16861-5G 16861-25G 16861-1G 16861-500MG
77631-776EA 841 USD
77631-776 77657-662 77640-890 77644-868
3-Bromo-2,6-dimethoxybenzaldehyde ≥97%
3-Bromo-2,6-dimethoxybenzaldehyde

Precaución: For Research Use Only

Formula: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD12025201
CAS Number: 29866-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR