4-Cyclopentyl-α,α-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

168136-5G 168136-25G 168136-1G 168136-500MG
77627-904EA 1530.58 USD
77627-904 77635-478 77640-886 77644-864
4-Cyclopentyl-α,α-dimethylbenzenemethanol ≥95%
4-Cyclopentyl-α,α-dimethylbenzenemethanol

Precaución: For Research Use Only

Formula: 204.31 g/mol
MW: C₁₄H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD26680022
CAS Number: 19920-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR