2-(Ethylthio)-4-fluoro-1-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

168102-5G 168102-500MG 168102-25G 168102-1G
77631-770EA 1186 USD
77631-770 77652-410 77653-810 77640-884
2-(Ethylthio)-4-fluoro-1-methylbenzene ≥95%
2-(Ethylthio)-4-fluoro-1-methylbenzene

Precaución: For Research Use Only

Formula: 170.25 g/mol
MW: C₉H₁₁FS
Storage Temperature: Ambient
MDL Number: MFCD11617739
CAS Number: 1314903-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR