1-(Ethylthio)-4-fluoro-2-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

168095-5G 168095-1G 168095-500MG
77657-660EA 1884 USD
77657-660 77640-882 77648-902
1-(Ethylthio)-4-fluoro-2-methoxybenzene ≥95%
1-(Ethylthio)-4-fluoro-2-methoxybenzene

Precaución: For Research Use Only

Formula: 186.25 g/mol
MW: C₉H₁₁FOS
Storage Temperature: Ambient
MDL Number: MFCD11617726
CAS Number: 1314937-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR