1-(Ethylthio)-2-(1-methylethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

168090-1G 168090-5G 168090-500MG
77636-886EA 630 USD
77636-886 77657-658 77644-862
1-(Ethylthio)-2-(1-methylethyl)benzene ≥95%
1-(Ethylthio)-2-(1-methylethyl)benzene

Precaución: For Research Use Only

Formula: 180.31 g/mol
MW: C₁₁H₁₆S
Storage Temperature: Ambient
MDL Number: MFCD00040058
CAS Number: 1314963-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR