1-(Ethylthio)-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

168076-5G 168076-1G 168076-25G 168076-500MG
77634-150EA 375 USD
77634-150 77636-882 77657-654 77644-860
1-(Ethylthio)-3-(trifluoromethyl)benzene ≥97%
1-(Ethylthio)-3-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 206.23 g/mol
MW: C₉H₉F₃S
Storage Temperature: Ambient
MDL Number: MFCD22484164
CAS Number: 1447671-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR