1-(Ethylthio)-3-fluoro-5-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

168068-5G 168068-25G 168068-1G 168068-500MG
77627-902EA 1484.23 USD
77627-902 77657-652 77640-880 77644-858
1-(Ethylthio)-3-fluoro-5-methoxybenzene ≥95%
1-(Ethylthio)-3-fluoro-5-methoxybenzene

Precaución: For Research Use Only

Formula: 186.25 g/mol
MW: C₉H₁₁FOS
Storage Temperature: Ambient
MDL Number: MFCD22483653
CAS Number: 1822762-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR