4-(Ethylthio)-1-fluoro-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

168066-5G 168066-25G 168066-1G 168066-500MG
77627-898EA 1484.23 USD
77627-898 77635-476 77636-878 77644-854
4-(Ethylthio)-1-fluoro-2-methoxybenzene ≥97%
4-(Ethylthio)-1-fluoro-2-methoxybenzene

Precaución: For Research Use Only

Formula: 186.25 g/mol
MW: C₉H₁₁FOS
Storage Temperature: Ambient
MDL Number: MFCD22483938
CAS Number: 1822764-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR